Product Details
- SNP ID
-
rs73158973
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:135557955 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTGCGCCTCTAAGATGGCGACGC[C/T]TTTGGCGGTAAATTCGGGTAAGTGT
- Phenotype
-
MIM: 614352
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP205
PubMed Links
Gene Details
- Gene
- NUP205
- Gene Name
- nucleoporin 205
View Full Product Details