Product Details

SNP ID
rs73694402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94127445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCCAAAGAGCAACATGTCACA[C/T]TGATTAAAGATGGTTAATGACACAG
Phenotype
MIM: 603017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH17 PubMed Links

Gene Details

Gene
CDH17
Gene Name
cadherin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144663.1 3420 UTR 3 NP_001138135.1
NM_004063.3 3420 UTR 3 NP_004054.3
XM_011516790.1 3420 UTR 3 XP_011515092.1

View Full Product Details