Product Details

SNP ID
rs73614641
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74713786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGACCCACTTCTGACTGTGAA[A/T]GCGAGCTGTGCCGTGCAGGCAGCTC
Phenotype
MIM: 611026
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FA2H PubMed Links

Gene Details

Gene
FA2H
Gene Name
fatty acid 2-hydroxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024306.4 1298 UTR 3 NP_077282.3
XM_011523317.2 1298 Intron XP_011521619.1
XM_011523319.2 1298 UTR 3 XP_011521621.1

View Full Product Details