Product Details
- SNP ID
-
rs73930688
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40036519 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGTCATTTCTAAAATAAAAGGCCT[C/T]GAGGCCAAGTGTTTTACTTATTTGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF780B
PubMed Links
Gene Details
- Gene
- ZNF780B
- Gene Name
- zinc finger protein 780B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005851.2 |
637 |
Missense Mutation |
AAG,GAG |
K114E |
NP_001005851.1 |
XM_005258590.4 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_005258647.1 |
XM_005258591.4 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_005258648.1 |
XM_005258592.3 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_005258649.1 |
XM_005258593.2 |
637 |
Missense Mutation |
AAG,GAG |
K114E |
XP_005258650.1 |
XM_005258595.3 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_005258652.1 |
XM_006723072.3 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_006723135.1 |
XM_011526593.2 |
637 |
Missense Mutation |
AAG,GAG |
K115E |
XP_011524895.1 |
XM_017026425.1 |
637 |
Missense Mutation |
AAG,GAG |
K114E |
XP_016881914.1 |
XM_017026426.1 |
637 |
Missense Mutation |
AAG,GAG |
K114E |
XP_016881915.1 |
XM_017026427.1 |
637 |
Missense Mutation |
AAG,GAG |
K114E |
XP_016881916.1 |
View Full Product Details