Product Details
- SNP ID
-
rs73382365
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:21670598 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAACAAATTTTCGTCGTTTACCT[C/T]TTGACCACTGCAGGTAAGAGTTTTG
- Phenotype
-
MIM: 607588
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC107985532
PubMed Links
Gene Details
- Gene
- LOC107985532
- Gene Name
- uncharacterized LOC107985532
There are no transcripts associated with this gene.
- Gene
- PPIL2
- Gene Name
- peptidylprolyl isomerase like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317996.1 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
NP_001304925.1 |
NM_014337.3 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
NP_055152.1 |
NM_148175.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
NP_680480.1 |
NM_148176.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
NP_680481.1 |
XM_005261448.3 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_005261505.1 |
XM_011530041.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528343.1 |
XM_011530042.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528344.1 |
XM_011530043.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528345.1 |
XM_011530044.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528346.1 |
XM_011530046.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528348.1 |
XM_011530047.2 |
231 |
Missense Mutation |
CTT,TTT |
L39F |
XP_011528349.1 |
XM_011530049.2 |
231 |
UTR 5 |
|
|
XP_011528351.1 |
XM_011530050.2 |
231 |
UTR 5 |
|
|
XP_011528352.1 |
XM_011530051.2 |
231 |
UTR 5 |
|
|
XP_011528353.1 |
XM_017028705.1 |
231 |
Intron |
|
|
XP_016884194.1 |
XM_017028706.1 |
231 |
Intron |
|
|
XP_016884195.1 |
View Full Product Details