Product Details

SNP ID
rs73266356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:112100243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAGCACTTTCAGTGCAGTGGTG[A/G]GAGCAGACATGAGATTGCATCTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC16 PubMed Links

Gene Details

Gene
ZCCHC16
Gene Name
zinc finger CCHC-type containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004308.2 Intron NP_001004308.2

View Full Product Details