Product Details

SNP ID
rs74111437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:80799227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGAAGAACCAGCTGCAACAAA[C/T]GTTAGTATTGCCCTCAGTCATGATT
Phenotype
MIM: 603380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIN7A PubMed Links

Gene Details

Gene
LIN7A
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324423.1 Intron NP_001311352.1
NM_004664.3 Intron NP_004655.1
XM_011538928.2 Intron XP_011537230.1

View Full Product Details