Product Details

SNP ID
rs73063677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46726761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATAATAGCATCCAGCTTTGTA[C/T]TGTTTACAAGAGATACACCTAAAAC
Phenotype
MIM: 607950
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRSS46 PubMed Links

Gene Details

Gene
PRSS46
Gene Name
protease, serine 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205271.1 Intron NP_001192200.1
Gene
PRSS50
Gene Name
protease, serine 50
There are no transcripts associated with this gene.

View Full Product Details