Product Details

SNP ID
rs74058052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63686133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATACCAATAAAGAAAAATATG[A/T]AAGGAACAAAAAATGTGATGGAGAA
Phenotype
MIM: 612826
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SGPP1 PubMed Links

Gene Details

Gene
SGPP1
Gene Name
sphingosine-1-phosphate phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030791.3 1449 Missense Mutation TAC,TTC Y433F NP_110418.1
XM_017021678.1 1449 UTR 3 XP_016877167.1

View Full Product Details