Product Details

SNP ID
rs73265491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:115828297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTATACGAACAAACTGAAGTTT[A/G]TACTTACGTTTCAAGGTTTTTGATA
Phenotype
MIM: 609608
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG12 PubMed Links

Gene Details

Gene
ATG12
Gene Name
autophagy related 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277783.1 4092 UTR 3 NP_001264712.1
NM_004707.3 4092 UTR 3 NP_004698.3

View Full Product Details