Product Details

SNP ID
rs75006481
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:158841639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTAGAGTCCATTCCACAACTCA[A/G]AACTGTAGTGCTGTGGCAGAAGGTT
Phenotype
MIM: 159553
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MNDA PubMed Links

Gene Details

Gene
MNDA
Gene Name
myeloid cell nuclear differentiation antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002432.1 Intron NP_002423.1
Gene
OR6N1
Gene Name
olfactory receptor family 6 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005185.1 Intron NP_001005185.1
XM_017000325.1 Intron XP_016855814.1
XM_017000326.1 Intron XP_016855815.1
XM_017000327.1 Intron XP_016855816.1

View Full Product Details