Product Details

SNP ID
rs78724201
Assay Type
Functionally Tested
NCBI dbSNP Submissions
10
Location
Chr.1:160219425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCCAATGAACCCCCAGAGAA[A/G]AGTCGGAGAACTATATTCATTTGGC
Phenotype
MIM: 615820 MIM: 603434
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DCAF8 PubMed Links

Gene Details

Gene
DCAF8
Gene Name
DDB1 and CUL4 associated factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015726.3 Intron NP_056541.2
Gene
PEA15
Gene Name
phosphoprotein enriched in astrocytes 15
There are no transcripts associated with this gene.

View Full Product Details