Product Details

SNP ID
rs77112568
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:172532566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACAGGGAAAGGGCTTGGTGCAG[C/T]TTCCCTCACAACACACACGTCATTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SUCO PubMed Links

Gene Details

Gene
SUCO
Gene Name
SUN domain containing ossification factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282750.1 218 Intron NP_001269679.1
NM_001282751.1 218 Intron NP_001269680.1
NM_014283.4 218 Intron NP_055098.1
NM_016227.3 218 Missense Mutation CTT,TTT L32F NP_057311.3
XM_006711374.2 218 Intron XP_006711437.1
XM_006711375.2 218 Intron XP_006711438.1
XM_006711376.1 218 Intron XP_006711439.1

View Full Product Details