Product Details

SNP ID
rs79743050
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:203168914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCTCAAACTCTCACCTTTGACC[C/T]CCAGCCGGCAGTCCACAGATGCCTC
Phenotype
MIM: 102775 MIM: 160795
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA1 PubMed Links

Gene Details

Gene
ADORA1
Gene Name
adenosine A1 receptor
There are no transcripts associated with this gene.

Gene
MYBPH
Gene Name
myosin binding protein H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004997.2 1468 Missense Mutation GAG,GGG E470G NP_004988.2

View Full Product Details