Product Details

SNP ID
rs409445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8380061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAAGGTTAAAAAAAAGAGCCCA[C/G]TTCGGCAACCTGTAAGAAGTTGACA
Phenotype
MIM: 603704
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRBA2 PubMed Links

Gene Details

Gene
KRBA2
Gene Name
KRAB-A domain containing 2
There are no transcripts associated with this gene.

Gene
RPL26
Gene Name
ribosomal protein L26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000987.3 Intron NP_000978.1
NM_001315530.1 Intron NP_001302459.1
NM_001315531.1 Intron NP_001302460.1

View Full Product Details