Product Details

SNP ID
rs177985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3862305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTGTTCAGGGGACACCAGGGA[C/G]AGAGGGACCTCAGGGTCAGAATATG
Phenotype
MIM: 611411
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAMKK1 PubMed Links

Gene Details

Gene
CAMKK1
Gene Name
calcium/calmodulin dependent protein kinase kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032294.2 Intron NP_115670.1
NM_172206.1 Intron NP_757343.1
NM_172207.2 Intron NP_757344.2
XM_005256817.1 Intron XP_005256874.1
XM_006721588.2 Intron XP_006721651.1
XM_011524031.2 Intron XP_011522333.2
XM_011524032.2 Intron XP_011522334.1
XM_011524033.1 Intron XP_011522335.1
XM_011524035.2 Intron XP_011522337.1
XM_017025219.1 Intron XP_016880708.1
XM_017025220.1 Intron XP_016880709.1
XM_017025221.1 Intron XP_016880710.1
XM_017025222.1 Intron XP_016880711.1

View Full Product Details