Product Details
- SNP ID
-
rs357854
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:49049124 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTTATTCTTCAAGCTGATCATTAT[C/T]TCCATAGAAGTTCTCCATAAAAATA
- Phenotype
-
MIM: 607461
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DYM
PubMed Links
Gene Details
- Gene
- DYM
- Gene Name
- dymeclin
- Gene
- MIR4744
- Gene Name
- microRNA 4744
There are no transcripts associated with this gene.
View Full Product Details