Product Details

SNP ID
rs35572315
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:112626968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAACTTTGCTTTGTGGATTCTTT[C/G]TTTTACATCTGGGTAACATTTAAGT
Phenotype
MIM: 601580
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CAPZA1 PubMed Links

Gene Details

Gene
CAPZA1
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006135.2 Intron NP_006126.1
XM_011542225.2 Intron XP_011540527.1
XM_017002424.1 Intron XP_016857913.1
Gene
ST7L
Gene Name
suppression of tumorigenicity 7 like
There are no transcripts associated with this gene.

View Full Product Details