Product Details

SNP ID
rs1059850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49859332 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAAAATATTTGCCCATAGACTAT[A/G]TGGAATATTATCATTAAAGTGTGAA
Phenotype
MIM: 609123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 4794 UTR 3 NP_079113.2
XM_011522046.2 4794 UTR 3 XP_011520348.1
XM_011522047.2 4794 UTR 3 XP_011520349.1
XM_011522048.1 4794 UTR 3 XP_011520350.1
XM_011522049.2 4794 UTR 3 XP_011520351.1
XM_011522051.2 4794 UTR 3 XP_011520353.1
XM_011522052.2 4794 UTR 3 XP_011520354.1
XM_011522053.1 4794 UTR 3 XP_011520355.1
XM_011522056.2 4794 UTR 3 XP_011520358.2
XM_011522058.2 4794 UTR 3 XP_011520360.1
XM_011522059.1 4794 UTR 3 XP_011520361.1
XM_011522060.1 4794 UTR 3 XP_011520362.1
XM_011522061.1 4794 UTR 3 XP_011520363.1
XM_011522062.1 4794 UTR 3 XP_011520364.1
XM_011522063.1 4794 UTR 3 XP_011520365.1
XM_011522064.1 4794 UTR 3 XP_011520366.1
XM_011522069.2 4794 UTR 3 XP_011520371.1
XM_011522070.1 4794 UTR 3 XP_011520372.1
XM_017022587.1 4794 UTR 3 XP_016878076.1
XM_017022588.1 4794 UTR 3 XP_016878077.1
XM_017022589.1 4794 UTR 3 XP_016878078.1
XM_017022590.1 4794 UTR 3 XP_016878079.1
XM_017022591.1 4794 UTR 3 XP_016878080.1
XM_017022592.1 4794 UTR 3 XP_016878081.1
XM_017022593.1 4794 Intron XP_016878082.1
XM_017022594.1 4794 UTR 3 XP_016878083.1
XM_017022595.1 4794 UTR 3 XP_016878084.1
XM_017022596.1 4794 UTR 3 XP_016878085.1
XM_017022597.1 4794 Intron XP_016878086.1
XM_017022598.1 4794 Intron XP_016878087.1
XM_017022599.1 4794 Intron XP_016878088.1

View Full Product Details