Product Details

SNP ID
rs11999672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127622419 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAAGGTAAGCTATTAGCCCAGTT[A/G]ACACATTTGTCCTGAACTTTTCCCT
Phenotype
MIM: 602926
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STXBP1 PubMed Links

Gene Details

Gene
STXBP1
Gene Name
syntaxin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032221.3 Intron NP_001027392.1
NM_003165.3 Intron NP_003156.1

View Full Product Details