Product Details

SNP ID
rs3761218
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:3795528 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAATGGGCGTGATACTTAATTCGC[C/T]TCTCTGGTTGCTGACACCCTCCTCG
Phenotype
MIM: 116949 MIM: 117140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC25B PubMed Links

Gene Details

Gene
CDC25B
Gene Name
cell division cycle 25B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287516.1 Intron NP_001274445.1
NM_001287517.1 Intron NP_001274446.1
NM_001287518.1 Intron NP_001274447.1
NM_001287519.1 Intron NP_001274448.1
NM_001287520.1 Intron NP_001274449.1
NM_001287522.1 Intron NP_001274451.1
NM_001287524.1 Intron NP_001274453.1
NM_004358.4 Intron NP_004349.1
NM_021872.3 Intron NP_068658.1
NM_021873.3 Intron NP_068659.1
Gene
CENPB
Gene Name
centromere protein B
There are no transcripts associated with this gene.

View Full Product Details