Product Details
- SNP ID
-
rs3771590
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241496823 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGGGACCAGATGGCTTCCTCCCAGC[C/T]GAGGGTCATGGAAGCCACTCAATGG
- Phenotype
-
MIM: 602255
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FARP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35084808] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FARP2
- Gene Name
- FERM, ARH/RhoGEF and pleckstrin domain protein 2
There are no transcripts associated with this gene.
- Gene
- STK25
- Gene Name
- serine/threonine kinase 25
View Full Product Details