Product Details

SNP ID
rs2481930
Assay Type
Functionally tested
NCBI dbSNP Submissions
69
Location
Chr.1:24561429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACATCTTCTTCAACCTGAAAGTC[A/G]CTTTTTCCTCTGGATTTTAAAAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506985 PubMed Links

Gene Details

Gene
LOC100506985
Gene Name
uncharacterized LOC100506985
There are no transcripts associated with this gene.

Gene
NCMAP
Gene Name
non-compact myelin associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010980.4 Intron NP_001010980.1
XM_005245872.3 Intron XP_005245929.1
XM_011541463.2 Intron XP_011539765.1

View Full Product Details