Product Details

SNP ID
rs12668105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143063769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGGCAGGAGTTTCCCTGGGGC[A/G]AGGAATGGATTACTGGCTGGGATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6W1P PubMed Links

Gene Details

Gene
OR6W1P
Gene Name
olfactory receptor family 6 subfamily W member 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details