Product Details
- SNP ID
-
rs8081395
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:59724400 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAGCTTTGGCATCTTAATAACTGG[A/G]CTAGGCACAGTGTCTCAAGCCTGTA
- Phenotype
-
MIM: 611753
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
VMP1
PubMed Links
Gene Details
- Gene
- VMP1
- Gene Name
- vacuole membrane protein 1
There are no transcripts associated with this gene.
View Full Product Details