Product Details
- SNP ID
-
rs1513982
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:101844956 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATATAGCCAAGTTTTTTTGTTTAA[A/C]GATGATGCATTCTTTCATTTTTTGT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR4F13P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75372058] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR4F13P
- Gene Name
- olfactory receptor family 4 subfamily F member 13 pseudogene
There are no transcripts associated with this gene.
View Full Product Details