Product Details

SNP ID
rs6672782
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:231818248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTTTGCCCATGCTGTGAATGTA[C/G]ATTAGCTGCTGCTAGATCTTCCATG
Phenotype
MIM: 605210 MIM: 606271
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DISC1 PubMed Links

Gene Details

Gene
DISC1
Gene Name
disrupted in schizophrenia 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012957.1 Intron NP_001012975.1
NM_001012958.1 Intron NP_001012976.1
NM_001012959.1 Intron NP_001012977.1
NM_001164537.1 Intron NP_001158009.1
NM_001164538.1 Intron NP_001158010.1
NM_001164539.1 Intron NP_001158011.1
NM_001164540.1 Intron NP_001158012.1
NM_001164541.1 Intron NP_001158013.1
NM_001164542.1 Intron NP_001158014.1
NM_001164544.1 Intron NP_001158016.1
NM_001164545.1 Intron NP_001158017.1
NM_001164546.1 Intron NP_001158018.1
NM_001164547.1 Intron NP_001158019.1
NM_001164548.1 Intron NP_001158020.1
NM_001164549.1 Intron NP_001158021.1
NM_001164550.1 Intron NP_001158022.1
NM_001164551.1 Intron NP_001158023.1
NM_001164552.1 Intron NP_001158024.1
NM_001164553.1 Intron NP_001158025.1
NM_001164554.1 Intron NP_001158026.1
NM_001164555.1 Intron NP_001158027.1
NM_001164556.1 Intron NP_001158028.1
NM_018662.2 Intron NP_061132.2
Gene
DISC2
Gene Name
disrupted in schizophrenia 2 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TSNAX-DISC1
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details