Product Details

SNP ID
rs7912227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68419956 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAAGAGTACTATAAGTACGCAA[C/G]TGGCCATAAAGACTATACTACCGAC
Phenotype
MIM: 601810
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNA2 PubMed Links

Gene Details

Gene
DNA2
Gene Name
DNA replication helicase/nuclease 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080449.2 Intron NP_001073918.2
XM_006717680.3 Intron XP_006717743.1
XM_011539417.1 Intron XP_011537719.1
XM_017015799.1 Intron XP_016871288.1

View Full Product Details