Product Details

SNP ID
rs11901412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:23947567 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACATTTGTAGCAGCTCACTCAA[C/T]TGATATAAATAAATTTTAGTTGGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBXN2A PubMed Links

Gene Details

Gene
UBXN2A
Gene Name
UBX domain protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181713.3 Intron NP_859064.2
XM_005264166.4 Intron XP_005264223.1
XM_005264167.4 Intron XP_005264224.1
XM_005264168.4 Intron XP_005264225.1
XM_005264169.4 Intron XP_005264226.1
XM_011532633.2 Intron XP_011530935.1
XM_017003517.1 Intron XP_016859006.1
XM_017003518.1 Intron XP_016859007.1

View Full Product Details