Product Details

SNP ID
rs10984148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:95807000 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCACAAATGCATTTCAATTAAA[C/T]ATACCCAAATATTTGCTGAGGAAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00476 PubMed Links

Gene Details

Gene
LINC00476
Gene Name
long intergenic non-protein coding RNA 476
There are no transcripts associated with this gene.

View Full Product Details