Product Details

SNP ID
rs789230
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:128883715 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGGTTTTGGAATGGATGGAGGCAG[A/G]GATTCCTTTAAGTTGGCTGTCACTT
Phenotype
MIM: 611103
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ACAD9 PubMed Links

Gene Details

Gene
ACAD9
Gene Name
acyl-CoA dehydrogenase family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014049.4 Intron NP_054768.2
XM_017006264.1 Intron XP_016861753.1
XM_017006265.1 Intron XP_016861754.1

View Full Product Details