Product Details
- SNP ID
-
hCV1862255
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:145689780 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGACCACACAGGATATGGGGGAGG[T/G]TGAAGAATAGGCCACAAAGTGCTCC
- Phenotype
-
MIM: 603831
- Polymorphism
- T/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PDZK1
PubMed Links
Gene Details
- Gene
- PDZK1
- Gene Name
- PDZ domain containing 1
There are no transcripts associated with this gene.
View Full Product Details