Product Details

SNP ID
rs11780207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22623006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGTGGGTGTGCACTGGCCAG[A/G]CTTCACGGCTGCGACTACGGGCCTG
Phenotype
MIM: 606396 MIM: 607359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BIN3 PubMed Links

Gene Details

Gene
BIN3
Gene Name
bridging integrator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018688.4 Intron NP_061158.1
XM_005273573.3 Intron XP_005273630.1
XM_011544586.2 Intron XP_011542888.1
XM_011544587.1 Intron XP_011542889.1
Gene
CCAR2
Gene Name
cell cycle and apoptosis regulator 2
There are no transcripts associated with this gene.

View Full Product Details