Product Details
- SNP ID
-
hCV1918276
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:20138917 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTATTGGATTAATTTTTTGTAAAGT[C/G]ATGTTTAAATTATATTCATAGTTAT
- Phenotype
-
MIM: 121015
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GJA3
PubMed Links
Gene Details
- Gene
- GJA3
- Gene Name
- gap junction protein alpha 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021954.3 |
4544 |
UTR 3 |
|
|
NP_068773.2 |
View Full Product Details