Product Details

SNP ID
rs3739435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22107602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTTCATGCCACCCCTCTGCTCCA[C/T]AGGGTCGAGGCCACAGGCAGTGACA
Phenotype
MIM: 602302 MIM: 615791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM160B2 PubMed Links

Gene Details

Gene
FAM160B2
Gene Name
family with sequence similarity 160 member B2
There are no transcripts associated with this gene.

Gene
HR
Gene Name
hair growth associated
There are no transcripts associated with this gene.

Gene
NUDT18
Gene Name
nudix hydrolase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024815.3 1520 Missense Mutation ATG,GTG M224V NP_079091.3
XM_011544650.1 1520 Missense Mutation ATG,GTG M440V XP_011542952.1

View Full Product Details