Product Details
- SNP ID
-
rs10733993
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29294502 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGGCAGAGCTCAGTCAAAGGCAT[C/T]TTCTCTGTGCTGCCTCTAGAGTGTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL1
PubMed Links
Gene Details
- Gene
- LYZL1
- Gene Name
- lysozyme like 1
View Full Product Details