Product Details

SNP ID
rs11535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232878822 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTAAAACACCTGGGCTCCTTAAG[C/G]GGCTAGAAGACACAAGTTACATCCA
Phenotype
MIM: 605991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C2orf82 PubMed Links

Gene Details

Gene
C2orf82
Gene Name
chromosome 2 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206895.1 2051 Intron NP_996778.1
XM_017004080.1 2051 Intron XP_016859569.1
XM_017004081.1 2051 Intron XP_016859570.1
XM_017004082.1 2051 Intron XP_016859571.1
XM_017004083.1 2051 Intron XP_016859572.1
XM_017004084.1 2051 Intron XP_016859573.1
Gene
NGEF
Gene Name
neuronal guanine nucleotide exchange factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114090.1 2051 UTR 3 NP_001107562.1
NM_019850.2 2051 UTR 3 NP_062824.2
XM_011510923.2 2051 UTR 3 XP_011509225.1
XM_011510924.1 2051 UTR 3 XP_011509226.1
XM_011510925.1 2051 UTR 3 XP_011509227.1

View Full Product Details