Product Details

SNP ID
rs1127390
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:80976176 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCCTGCTCTTGGTCCTGTTTTCTA[C/T]GTACTGAAAAATAAAAGAAGGGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CMC2 PubMed Links

Gene Details

Gene
CMC2
Gene Name
C-X9-C motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020188.3 439 Missense Mutation ATA,GTA I53V NP_064573.1
XM_017023461.1 439 Missense Mutation ATA,GTA I179V XP_016878950.1
XM_017023462.1 439 UTR 3 XP_016878951.1
XM_017023463.1 439 Intron XP_016878952.1
XM_017023464.1 439 Missense Mutation ATA,GTA I136V XP_016878953.1
XM_017023465.1 439 UTR 3 XP_016878954.1
XM_017023466.1 439 Missense Mutation CAT,CGT H97R XP_016878955.1
XM_017023467.1 439 Missense Mutation ATA,GTA I53V XP_016878956.1
XM_017023468.1 439 Missense Mutation ATA,GTA I53V XP_016878957.1
XM_017023469.1 439 Missense Mutation ATA,GTA I53V XP_016878958.1
XM_017023470.1 439 Missense Mutation ATA,GTA I53V XP_016878959.1

View Full Product Details