Product Details

SNP ID
rs732360
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:44222379 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GACGTTTCCACTCTTTGCTGTGAGT[A/G]CTGCCTCCAACGGCCACATACATGC
Phenotype
MIM: 607707 MIM: 606209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAMK2B PubMed Links

Gene Details

Gene
CAMK2B
Gene Name
calcium/calmodulin dependent protein kinase II beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220.4 Intron NP_001211.3
NM_001293170.1 Intron NP_001280099.1
NM_172078.2 Intron NP_742075.1
NM_172079.2 Intron NP_742076.1
NM_172080.2 Intron NP_742077.1
NM_172081.2 Intron NP_742078.1
NM_172082.2 Intron NP_742079.1
NM_172083.2 Intron NP_742080.1
NM_172084.2 Intron NP_742081.1
XM_005249862.3 Intron XP_005249919.1
XM_005249864.3 Intron XP_005249921.1
XM_006715781.2 Intron XP_006715844.1
XM_006715784.2 Intron XP_006715847.1
XM_011515547.1 Intron XP_011513849.1
XM_011515549.1 Intron XP_011513851.1
XM_011515550.1 Intron XP_011513852.1
XM_011515551.1 Intron XP_011513853.1
XM_011515552.1 Intron XP_011513854.1
XM_011515553.1 Intron XP_011513855.1
XM_011515554.1 Intron XP_011513856.1
XM_011515555.1 Intron XP_011513857.1
XM_011515556.1 Intron XP_011513858.1
XM_011515557.1 Intron XP_011513859.1
XM_011515558.2 Intron XP_011513860.1
XM_011515559.1 Intron XP_011513861.1
XM_017012660.1 Intron XP_016868149.1
XM_017012661.1 Intron XP_016868150.1
XM_017012662.1 Intron XP_016868151.1
XM_017012663.1 Intron XP_016868152.1
XM_017012664.1 Intron XP_016868153.1
XM_017012665.1 Intron XP_016868154.1
XM_017012666.1 Intron XP_016868155.1
XM_017012667.1 Intron XP_016868156.1
XM_017012668.1 Intron XP_016868157.1
XM_017012669.1 Intron XP_016868158.1
XM_017012670.1 Intron XP_016868159.1
Gene
YKT6
Gene Name
YKT6 v-SNARE homolog (S. cerevisiae)
There are no transcripts associated with this gene.

View Full Product Details