Product Details

SNP ID
hCV2327341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:50668292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCCTCGGTTTTAAATAGCAGGA[A/G]GAGGGGAATCAATGTTTGATTCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARP8 PubMed Links

Gene Details

Gene
PARP8
Gene Name
poly(ADP-ribose) polymerase family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178055.1 Intron NP_001171526.1
NM_001178056.1 Intron NP_001171527.1
NM_024615.3 Intron NP_078891.2

View Full Product Details