Product Details

SNP ID
rs3851729
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:74451631 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTTTTAGGCACGTGATTTATAATAC[C/T]TTGTGGACATCTAACAGGTTAATAA
Phenotype
MIM: 600753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLG1 PubMed Links

Gene Details

Gene
GLG1
Gene Name
golgi glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145666.1 4054 UTR 3 NP_001139138.1
NM_001145667.1 4054 UTR 3 NP_001139139.1
NM_012201.5 4054 UTR 3 NP_036333.2

View Full Product Details