Product Details

SNP ID
rs4904448
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:88385822 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCGGCTGCAAGAGGACTAAGCATG[A/G]ATGGCAGCCGGAGAGGTAAAGGGCA
Phenotype
MIM: 609868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPATA7 PubMed Links

Gene Details

Gene
SPATA7
Gene Name
spermatogenesis associated 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040428.3 150 Missense Mutation AAT,GAT N2D NP_001035518.1
NM_018418.4 150 Missense Mutation AAT,GAT N2D NP_060888.2
XM_005267851.1 150 Missense Mutation AAT,GAT N2D XP_005267908.1
XM_005267852.1 150 Missense Mutation AAT,GAT N2D XP_005267909.1
XM_005267854.1 150 UTR 5 XP_005267911.1
XM_005267855.1 150 UTR 5 XP_005267912.1
XM_006720204.1 150 Missense Mutation AAT,GAT N2D XP_006720267.1
XM_006720205.1 150 Missense Mutation AAT,GAT N2D XP_006720268.1
XM_011536951.1 150 UTR 5 XP_011535253.1
XM_011536952.1 150 Missense Mutation AAT,GAT N2D XP_011535254.1
XM_011536953.1 150 UTR 5 XP_011535255.1
XM_017021452.1 150 UTR 5 XP_016876941.1
XM_017021453.1 150 UTR 5 XP_016876942.1
XM_017021454.1 150 UTR 5 XP_016876943.1
XM_017021455.1 150 UTR 5 XP_016876944.1
XM_017021456.1 150 Intron XP_016876945.1
XM_017021457.1 150 UTR 5 XP_016876946.1

View Full Product Details