Product Details

SNP ID
rs6073834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45651350 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACATCCAGTACAAATCGTACTTT[A/C]AGTACCCATCAACCATTCTGGAGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WFDC11 PubMed Links

Gene Details

Gene
WFDC11
Gene Name
WAP four-disulfide core domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147197.2 Intron NP_671730.1

View Full Product Details