Product Details
- SNP ID
-
rs1807591
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:42798267 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCCTGGGGCTGAGGGAACTCTCCG[A/G]AATGGGTGCACCTCCACGCCCTCCT
- Phenotype
-
MIM: 612439
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARFGAP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116982580] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARFGAP3
- Gene Name
- ADP ribosylation factor GTPase activating protein 3
View Full Product Details