Product Details

SNP ID
rs2106107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29439998 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTGCAGACGCTGGTCTCCTCAT[C/T]GGTGTGCCATGGACTTCAACAGGGG
Phenotype
MIM: 605968 MIM: 605972
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RFPL1 PubMed Links

Gene Details

Gene
RFPL1
Gene Name
ret finger protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021026.2 Intron NP_066306.2
XM_011530298.2 Intron XP_011528600.1
XM_011530299.2 Intron XP_011528601.1
XM_017028900.1 Intron XP_016884389.1
XM_017028901.1 Intron XP_016884390.1
XM_017028902.1 Intron XP_016884391.1
Gene
RFPL1S
Gene Name
RFPL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details