Product Details

SNP ID
rs1045953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120426652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTGAATGTATCTGAATTTCCA[A/G]TTTATATGCTTAGCAAGGTCACAAG
Phenotype
MIM: 309060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMP2 PubMed Links

Gene Details

Gene
LAMP2
Gene Name
lysosomal associated membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122606.1 6084 UTR 3 NP_001116078.1
NM_002294.2 6084 UTR 3 NP_002285.1
NM_013995.2 6084 Intron NP_054701.1
Gene
LOC105373326
Gene Name
uncharacterized LOC105373326
There are no transcripts associated with this gene.

View Full Product Details