Product Details

SNP ID
rs10488898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:106922336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTCTACAAAAGTTTAAAGTCAC[A/G]ACTATAAGATAACCTTTTATTGTTT
Phenotype
MIM: 605415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKK2 PubMed Links

Gene Details

Gene
DKK2
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014421.2 3103 UTR 3 NP_055236.1

View Full Product Details