Product Details

SNP ID
rs419764
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:106923000 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTACCAATCTAATTCTATCAAAATA[C/T]AGGTTATGGCCAATCTTTAAGAAAT
Phenotype
MIM: 605415
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DKK2 PubMed Links

Gene Details

Gene
DKK2
Gene Name
dickkopf WNT signaling pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014421.2 2439 UTR 3 NP_055236.1

View Full Product Details