Product Details

SNP ID
rs2074520
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:35012264 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGATAACAGGTACATTGTAAACAAA[C/G]TCTCAGTGCACATGCTTCCCTCAAA
Phenotype
MIM: 600940 MIM: 609735
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LIG3 PubMed Links

Gene Details

Gene
LIG3
Gene Name
DNA ligase 3
There are no transcripts associated with this gene.

Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

Gene
RFFL
Gene Name
ring finger and FYVE-like domain containing E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017368.1 Intron NP_001017368.1

View Full Product Details