Product Details

SNP ID
rs11973275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26163235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCTTTCTTACTCTTCCACTACT[A/T]CCATTAACTCAAAACCGTCTGTAAA
Phenotype
MIM: 604135
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NFE2L3 PubMed Links

Gene Details

Gene
NFE2L3
Gene Name
nuclear factor, erythroid 2 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004289.6 Intron NP_004280.5

View Full Product Details